Learn More
Zeiss™ Objective LD A-Plan
Designed for routine research. A-Plan Objectives are ideal for a variety of applications.
788.00€ - 1733.10€
Product Code | Brand | Magnification Power | Focus Distance | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Magnification Power | Focus Distance | Price | Quantity & Availability | ||||
14811251
|
Zeiss™
4212419910 |
10X | 8.5 mm |
1155.00€
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
14801251
|
Zeiss™
4212319910 |
20X | 4.3 mm |
834.00€
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
12113921
|
Zeiss™
4210909902 |
100X | 0.22 mm |
1005.00€
Each |
Estimated Shipment: 10-05-2024
Log in to see stock. |
||||
13264369
|
Zeiss™
421090-9902-000 |
100X | 0.22 mm |
788.00€
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
11855163
|
Zeiss™
4212519910 |
20X | 4.3 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
11875163
|
Zeiss™
4212619916 |
40X | 2 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
11865163
|
Zeiss™
4212619910 |
32X | 3.1 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
12163931
|
Zeiss™
421261-9910-000 |
32X | 3.1 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|||||
Description
- This class of objectives provides an attractive alternative to Achroplan objectives, both in routine and research applications
- A-Plan objectives provide rich contrast and can be used with eyepieces with a field of view up to 23mm
- Allow contrasting methods ranging from brightfield to phase contrast
- For excitation wavelength within the visible spectrum they can be used for epifluorescence work
Specifications
Objective LD A-Plan | |
1 Ea. |