Learn More
Zeiss™ Objective LD A-Plan
Designed for routine research. A-Plan Objectives are ideal for a variety of applications.
Brand: Zeiss™ 4210909902
Additional Details : Weight : 0.80000kg
Description
- This class of objectives provides an attractive alternative to Achroplan objectives, both in routine and research applications
- A-Plan objectives provide rich contrast and can be used with eyepieces with a field of view up to 23mm
- Allow contrasting methods ranging from brightfield to phase contrast
- For excitation wavelength within the visible spectrum they can be used for epifluorescence work
Specifications
Objective LD A-Plan | |
0.22 mm | |
100X |
Objective A-Plan 100x/1.25 Oil M27 | |
Immersol 518 N, oiler 20mL | |
1 Ea. |