Learn More
Zeiss™ Objective LD A-Plan
Click to view available options
Magnification Power:
100X
10X
20X
40X
Focus Distance:
0.22 mm
2 mm
4.3 mm
8.5 mm
Unit Size:
Each
Description
- This class of objectives provides an attractive alternative to Achroplan objectives, both in routine and research applications
- A-Plan objectives provide rich contrast and can be used with eyepieces with a field of view up to 23mm
- Allow contrasting methods ranging from brightfield to phase contrast
- For excitation wavelength within the visible spectrum they can be used for epifluorescence work
Specifications
Specifications
| Type | Objective LD A-Plan |
| Focus Distance | 2 mm |
| Magnification Power | 40X |
| Quantity | 1 Ea. |