Learn More
Carl Zeiss™ Objective EC Plan Neofluar
Click to view available options
Magnification Power:
100X
20X
40X
Unit Size:
Each
Description
- Enhanced Contrast Plan-Neofluar
- Minimized stray light: with a visible increase in contrast important
- High transmission of wavelengths from the near UV range (>40% starting at 340nm)
- Glass types of low autofluorescence makes these objectives
- Ideal for epi-fluorescence microscopy
Specifications
Specifications
| Type | Objective EC |
| Magnification Power | 100X |
| Quantity | 1 Ea. |