Learn More
Zeiss™ N-Achroplan Objectives
Designed for transmitted-light routine microscopy and epi-fluorescence microscopy with visible-light excitation. N-Achroplan Objectives are solid and reliable, ideal for photomicrography in routine microscopy.
Brand:  Zeiss™ 420980-9900-000
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11855023
  • 2660.60€ / Each
Estimated Shipment: 15-07-2024
Description
  • Excellent image flatness across visual field diameters of 23mm
  • Ideal for photomicrography in routine microscopy
  • Excellent flatness of field
  • Highly recommendable solution for image documentation in pathology
  • Well-suited for microphotography
  • Available in various versions for a wide range of applications
  • Possible contrast techniques include brightfield and phase contrast in transmitted light and fluorescence with excitation in the visible range
Mechanical correction collar for:
  • Cover glass thickness correction
  • Different immersion
  • Different temperature
  • Adjusting an iris diaphragm
Specifications
Specifications
Objective
63X