Learn More
Zeiss™ N-Achroplan Objectives
Designed for transmitted-light routine microscopy and epi-fluorescence microscopy with visible-light excitation. N-Achroplan Objectives are solid and reliable, ideal for photomicrography in routine microscopy.
2660.60€
Specifications
Type Objective
Quantity 1 Ea.
View More Specs
Products 2
Product Code Brand Magnification Power Price Quantity & Availability  
Product Code Brand Magnification Power Price Quantity & Availability  
11822300
View Documents Promotion Details
Zeiss™
420970-9900-000
50X
2660.60€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
11855023
View Documents Promotion Details
Zeiss™
420980-9900-000
63X
2660.60€
Each
View alternative products
Save Up to 
»
Estimated Shipment: 02-07-2024
Description
  • Excellent image flatness across visual field diameters of 23mm
  • Ideal for photomicrography in routine microscopy
  • Excellent flatness of field
  • Highly recommendable solution for image documentation in pathology
  • Well-suited for microphotography
  • Available in various versions for a wide range of applications
  • Possible contrast techniques include brightfield and phase contrast in transmitted light and fluorescence with excitation in the visible range
Mechanical correction collar for:
  • Cover glass thickness correction
  • Different immersion
  • Different temperature
  • Adjusting an iris diaphragm
Specifications
Specifications
Objective