Learn More
Zeiss™ N-Achroplan Objectives
Designed for transmitted-light routine microscopy and epi-fluorescence microscopy with visible-light excitation. N-Achroplan Objectives are solid and reliable, ideal for photomicrography in routine microscopy.
Brand: Zeiss™ 420970-9900-000
Additional Details : Weight : 0.80000kg
Description
- Excellent image flatness across visual field diameters of 23mm
- Ideal for photomicrography in routine microscopy
- Excellent flatness of field
- Highly recommendable solution for image documentation in pathology
- Well-suited for microphotography
- Available in various versions for a wide range of applications
- Possible contrast techniques include brightfield and phase contrast in transmitted light and fluorescence with excitation in the visible range
Mechanical correction collar for:
- Cover glass thickness correction
- Different immersion
- Different temperature
- Adjusting an iris diaphragm
Specifications
Objective | |
50X | |
1 Ea. |