Learn More
Carl Zeiss™ Objective A Achromat
Product Code. 13502340
Change view
Click to view available options
Type:
M27
M27, 23mm Field of View
Objective A
Oil PH3 M27
PH2 M27
Ph1 M27
Magnification Power:
100X
10X
2.5X
20X
40X
5X
Numerical Aperture:
0.12 mm
0.25 mm
0.45 mm
0.65 mm
1.25 mm
0.06 m
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Magnification Power Numerical Aperture unitSize
13502340 Oil PH3 M27 100X 1.25 mm Each
13469128 Oil PH3 M27 100X 1.25 mm Each
11832290 Objective A 10X 0.25 mm Each
11845163 Ph1 M27 10X 0.25 mm Each
11805063 Ph1 M27 10X 0.25 mm Each
11855083 M27 40X 0.65 mm Each
11845023 M27 2.5X 0.06 m Each
11815063 PH2 M27 20X 0.45 mm Each
11825063 M27, 23mm Field of View 40X 0.65 mm Each
11895063 M27 20X 0.45 mm Each
11895053 M27 5X 0.12 mm Each
11825163 M27 5X 0.12 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 13502340 Supplier Carl Zeiss™ Supplier No. 4210919912
This item requires a price update prior to being ordered.
Request a Price for this item here.
Use these universal objectives for routine research. Zeiss™ A-Plan Objectives provide rich contrast and can be used with eyepieces with a field of view up to 23mm.
  • This class of objectives provides an attractive alternative to Achroplan objectives, both in routine and research applications
  • Allow contrasting methods ranging from brightfield to phase contrast and for excitation wavelength within the visible spectrum
  • Can be used for epifluorescence work
Specifications
Type Oil PH3 M27
Description Oil PH3 M27
Magnification Power 100X
Numerical Aperture 1.25 mm
Quantity 1 Ea.