Learn More
Zeiss™ Objective
Description
For use with a variety of applications
Specifications
Specifications
| Type | Objective N-Achroplan 5x/0.15 Ph1 M27 |
| Description | Numerical Aperture: 0.15 |
| Focus Distance | 12 mm |
| For Use With (Equipment) | Axio Imager, Axio Lab, Axio Observer, Axio Vert and Axio Scope Microscopes |
| Magnification Power | 5X |
| Quantity | 1 Ea. |