Learn More
buffer pouches, Cole-Parmer Instrument Company, Oakton™
Economical pouches are accurate and convenient.
Brand:  Thermo Scientific Eutech 35653-02_CLEAN
Product Code. 12961678
  • ONLINE EXCLUSIVE PRICE
    93.75€
    62.10€ / 20mL
    Save 31.65€
    33% Off
Description
Applications: Electrochemistry, Laboratory Research, General Research, Pharmaceutical