Learn More
Oakton™ Testr PCTS 50 Waterproof Pocket pH/Cond/TDS/Salinity Tester
Product Code. 15949511
Change view
Click to view available options
Range (pH):
-1 to 15
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range (pH) unitSize
15949511 -1 to 15 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15949511 Supplier Oakton™ Supplier No. 3563435
Only null left
Add to basket
Add to basket
Includes
Protective plastic storage case, lanyard, and batteries.
Great for water, wastewater, laboratory or plant use, these multiparameter testers feature long-life pH electrodes with a wide range of sample compatibility. Ideal for pH, conductivity, TDS, salinity, and temperature testing.
  • Combine measurement capabilities of three testers in one convenient pocket-sized unit
  • Up to 3 point pH calibration with NIST or USA auto buffer recognition
  • Conductivity, TDS, and salinity feature auto ranging based on the concentration of your sample
  • Redesigned leakproof cap functions as a sample/calibration cup, hydrating cup, and stabilizing base to stand the meter upright
Warranty
1 Year
Specifications
Accuracy (Conductivity) ±1% full scale
Accuracy (pH) ±0.01
Accuracy (TDS) ±1% full scale
Backlight Option Yes
Battery Life 150+ hours
Battery Type 4 AAA Alkaline
Calibration Up to 3 points
Calibration Points 3
Channels 1
Display Type Graphics, Dot Matrix 80 x 100 pixel
IP Rating IP67
Measurement Modes pH, Conductivity, TDS, Salinity, Temperature
Range (Conductivity) 0 to 20.00 mS
Range (pH) -1 to 15
Range (Salinity) 0 to 10.00 ppt
Range (TDS) 0 to 10.00 ppt
Resolution (pH) 0.01
Temperature Range (Metric) 0°C to 60°C
For Use With (Application) Water quality testing, buffer and reagent production, electrochemistry
Includes Protective plastic storage case, lanyard, and 4 AAA batteries.
Keypad Membrane
Languages English
Memory Hold Data
Model 50 Series
Probe Type Integral multiparameter probe
Resolution Salinity: 0.10 ppt
Resolution (Conductivity) 0.1 μS, 1 μS; 0.01 mS
Resolution (TDS) 0.1 ppm, 1 ppm, 0.01 ppt
Stability Checkmark indicator
Temperature Accuracy ±0.9°F (0.5°C) from 32 to 122°F (0 to 50°C); ±1.8°F (1.0°C) from 122 to 140°F (50 to 60°C)
Temperature Compensation Automatic
Temperature (English) Operating 41°F to 113°F (ambient)
Temperature (Metric) Operating 5°C to 45°C
Temperature Range (English) 32°F to 140°F
Temperature Resolution 0.1°F/°C
Warranty 1 Year