Learn More
Digi-Sense™ Infrared Food Safety Thermometers
Product Code. p-8004329 Shop All Cole-Parmer Products
Click to view available options
Temperature Range (Metric):
-18°C to +302°C
-30°C to +200°C
-35°C to +275°C
Unit Size:
Each
Product Code. 11781485
Brand: Digi-Sense™ 3562545
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ideal for food inspections and HACCP programs.
  • Can also be used in the home, office, field or lab, anywhere you need to quickly and accurately measure surface temperatures
Specifications
Product Type Food Infrared Thermometer
Max. Temperature (Metric) 200°C
Min. Temperature (Metric) -30°C
Temperature Range (Metric) -30°C to +200°C