Learn More
Digi-Sense™ Temp 10 Series Thermocouple thermometer
Product Code. p-203772 Shop All Cole-Parmer Products
Click to view available options
Temperature Range (Metric):
-210°C to +1200°C
Temperature Range (English):
-346°F to +2192°F
Probe Type:
Type J Thermocouple
Unit Size:
Each
Product Code. 16614954
Brand: Digi-Sense™ 9142800
This item requires a price update prior to being ordered.
Request a Price for this item here.
Includes
Three AA batteries provide 700 hours continuous (without use of backlighting)
Requires
Probes are sold separately
Heavy-duty thermometer offers versatility and accuracy in a compact design
  • Push-button operation
  • Ergonomic design with large easy-to-read screen with back light
  • Display either °C or °F
  • Minimum and maximum temperature display
  • Temperature offset calibration adjustment
  • Hold function – freezes measurements for convenient reading and recording
  • Accepts two blade female ANSI miniconnector input
  • Auto-off function – turns off meter after 17 minutes of nonuse to save batteries
  • Sealed keypad and ABS plastic case – Meet IP54 standards for splash resistance
Order Info
  • Available rubber armor – Protects meter; features a built-in stand
  • Optional 3-way hands-free kit
Certifications
UL listed and certified for Class I: Groups A, B, C, and D division, NIST™ traceable. (Cat. No. 13-621-855 comes with NIST certification.)
Specifications
Certifications/Compliance Not certified
Depth (English) 2 in.
Depth (Metric) 5.2 cm
Height (English) 7 in.
Height (Metric) 17.8 cm
Includes Three AA battery and instructions
Length (Metric) 175 mm
Material ABS Plastic
Probe Type Type J Thermocouple
Resolution 1°C
Weight (English) 20 oz.
Weight (Metric) 600 g
Width (English) 4 in.
Product Type Thermocouple Thermometer
Max. Temperature (English) 2192°F
Max. Temperature (Metric) 1200°C
Min. Temperature (English) -346°F
Min. Temperature (Metric) -210°C
Temperature Range (English) -346°F to +2192°F
Temperature Range (Metric) -210°C to +1200°C
Calibrated No
Battery Type AA
Accuracy ±0.4°C
Display Digital LCD
Width (Metric) 18 cm