Learn More
Aalborg™ Analog Thermal Mass Flowmeter for Oxygen
Product Code. 11738468
Click to view available options
Type:
Mass Flowmeter
Unit Size:
Each
Product Code. 11738468
Brand: Aalborg™ GFM47AVDL6A0/3OB40SS
Only null left
Add to basket
Add to basket
Designed to read flow rates of gases in a low cost and convenient manner. Aalborg™ Analog Thermal Mass Flowmeter for Oxygen has rugged design coupled with instrumentation grade accuracy to provide versatile and economical means of flow measurement.
  • Rigid metallic construction
  • Built-in tiltable LCD readout
  • Circuit protection
  • Can be used as portable device
  • Engineering units or 0 to 100% displays
  • TIO - Totalizer option
Specifications
Type Mass Flowmeter
Accuracy ±1.5%
Flow Rate Range 0 to 60 LPM