missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Tolylglyoxylic-d7 Acid Methyl Ester, TRC
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.5mg
5mg
Specifications
Specifications
| Chemical Name or Material | o-Tolylglyoxylic-d7 Acid Methyl Ester |
| Molecular Formula | C10 D7 H3 O3 |
| Quantity | 5 mg |
| Synonym | (2-Methylphenyl)glyoxylic-d7 Acid Methyl Ester, Methyl 2-(2-Methylphenyl)-2-oxoacetate-d7, Methyl 2-Methylphenylglyoxylate-d7, Methyl o-Toluoylformate-d7 |
| InChI Formula | InChI=1S/C10H10O3/c1-7-5-3-4-6-8(7)9(11)10(12)13-2/h3-6H,1-2H3/i1D3,3D,4D,5D,6D |
| SMILES | [2H]c1c([2H])c([2H])c(c(C(=O)C(=O)OC)c1[2H])C([2H])([2H])[2H] |
| IUPAC Name | methyl 2-oxo-2-[2,3,4,5-tetradeuterio-6-(trideuteriomethyl)phenyl]acetate |
| Molecular Weight (g/mol) | 185.228 |
| Formula Weight | 185.107 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction