missing translation for 'onlineSavingsMsg'
Learn More

o-Tolyl Cyanide, TRC

Product Code. 30453973
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453973 50 g 50g
30455413 1 g 1g
30455309 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453973 Supplier Toronto Research Chemicals Supplier No. TRCT53660550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 529-19-1
Molecular Formula C8 H7 N
Molecular Weight (g/mol) 117.15
Synonym Benzonitrile, 2-methyl- (9CI, ACI), 2-Methylbenzonitrile (ACI), o-Tolunitrile (8CI), 1-Cyano-2-methylbenzene, 2-Cyanotoluene, 2-Methylbenzenecarbonitrile, 2-Tolunitrile, 2-Tolyl cyanide, NSC 66549, o-Cyanotoluene, o-Methylbenzonitrile, o-Toluenecarbonitrile, o-Tolyl cyanide
IUPAC Name 2-methylbenzonitrile
SMILES Cc1ccccc1C#N

Specifications

Quantity 50 g
InChI Formula InChI=1S/C8H7N/c1-7-4-2-3-5-8(7)6-9/h2-5H,1H3
Formula Weight 117.0578
Chemical Name or Material o-Tolyl Cyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.