missing translation for 'onlineSavingsMsg'
Learn More

o-Toluic acid, 98+%

Product Code. 11419060
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
This item is not returnable. View return policy

Product Code. 11419060

Brand: Thermo Scientific Alfa Aesar A13856.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 118-90-1 | C8H8O2 | 136.15 g/mol

o-Toluic acid is used for producing of fluorescent whitening agent ER33%, bensulfuronmethyl,o-tolunitrile,o-cyano-benzaldehyde and 3,5-dinitro-o-toluylamide etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Toluic acid is used for producing of fluorescent whitening agent ER33%, bensulfuronmethyl,o-tolunitrile,o-cyano-benzaldehyde and 3,5-dinitro-o-toluylamide etc.

Solubility
Soluble in alcohol and chloroform. Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-90-1
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002477
InChI Key ZWLPBLYKEWSWPD-UHFFFAOYSA-N
Synonym o-toluic acid, o-toluylic acid, orthotoluic acid, 2-toluic acid, toluic acid, benzoic acid, 2-methyl, o-methylbenzoic acid, 2-methyl-benzoic acid, benzoic acid, methyl, o-toluate
PubChem CID 8373
ChEBI CHEBI:36632
IUPAC Name 2-methylbenzoic acid
SMILES CC1=CC=CC=C1C(=O)O

Specifications

Melting Point 103°C to 106°C
Density 1.06
Boiling Point 258°C to 259°C
Flash Point 148°C (298°F)
Odor Aromatic
Quantity 1000 g
Beilstein 1072103
Merck Index 14,9535
Solubility Information Soluble in alcohol and chloroform. Slightly soluble in water.
Formula Weight 136.15
Percent Purity ≥98%
Chemical Name or Material o-Toluic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.