missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Tolualdehyde-13C1 (carbonyl-13C), CDN
Click to view available options
Quantity:
0.5 g
Unit Size:
0.5g
Specifications
Specifications
| Chemical Name or Material | o-Tolualdehyde-13 C (carbonyl-13 C) |
| CAS | 138151-99-2 |
| Molecular Formula | 13 C C7 H8 O |
| Quantity | 0.5 g |
| Synonym | Benzaldehyde-formyl-13 C, 2-methyl- (9 CI), o-Tolualdehyde-13 C (carbonyl-13 C), 2-Methylbenzaldehyde-formyl-13 C |
| InChI Formula | InChI=1 S/C8H8O/c1-7-4-2-3-5-8(7)6-9/h2-6 H,1H3/i6+1 |
| SMILES | Cc1ccccc1[13 CH]=O |
| IUPAC Name | 2-methyl(^{13}C)benzaldehyde |
| Molecular Weight (g/mol) | 121.141 |
| Formula Weight | 121.061 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction