missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Tolidine, 95%, pract
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
Unit Size:
100g
250g
25g
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Specifications
Specifications
| Melting Point | 125.0°C to 132.0°C |
| Color | Beige or Brown to Gray |
| Boiling Point | 300.5°C |
| Flash Point | 244°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 95% |
| Packaging | Glass bottle |
| Molecular Formula | C14H16N2 |
| Quantity | 500 g |
| Beilstein | 13, IV, 419 |
| Show More |
Safety and Handling
Product Identifier
- o-Tolidine
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 1B
- H350-May cause cancer.
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction