Learn More
O-(Tetrahydro-2H-pyran-2-yl)hydroxylamine, TRC
Specifications
Specifications
| Chemical Name or Material | O-(Tetrahydro-2H-pyran-2-yl)hydroxylamine |
| CAS | 6723-30-4 |
| Molecular Formula | C5H11NO2 |
| Quantity | 500 mg |
| Synonym | 2-(Aminoxy)tetrahydropyran, 2-Tetrahydropyranyloxyamine, O-(Tetrahydropyran-2-yl)hydroxylamine, O-Tetrahydropyranylhydroxylamine, [(Tetrahydro-2H-pyranyl-2-yl)oxy]amine |
| InChI Formula | InChI=1S/C5H11NO2/c6-8-5-3-1-2-4-7-5/h5H,1-4,6H2 |
| SMILES | NOC1CCCCO1 |
| IUPAC Name | O-(oxan-2-yl)hydroxylamine |
| Molecular Weight (g/mol) | 117.15 |
| Formula Weight | 117.079 |
Product Title
Haben Sie Verbesserungsvorschläge?