Learn More
Merck O-Ring
Product Code. 10325431 Shop All Merck Products
Change view
Click to view available options
For Use With (Equipment):
25mm Gas Line Filter Holder
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10325431 25mm Gas Line Filter Holder Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10325431 Supplier Merck Supplier No. XX4002516
  • ONLINE EXCLUSIVE PRICE
    114.00€
    85.30€ / Pack of 6
    Save 28.70€
    25% Off
Estimated Shipment: 20-10-2026
Specifications
For Use With (Application) Viton 2-021
For Use With (Equipment) 25mm Gas Line Filter Holder
Quantity 5 Pack
Product Type O-Ring
Material Fluoroelastomer
Diameter (Metric) Outside 23.52 mm
Size 2-021