missing translation for 'onlineSavingsMsg'
Learn More

O,O-Diethyl Thiophosphate Sodium Salt, TRC

Product Code. 30371428
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30371428 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30371428 Supplier Toronto Research Chemicals Supplier No. TRCD4451252.5G
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O,O-Diethyl Thiophosphate Sodium Salt
CAS 5852-63-1
Molecular Formula C4H10NaO3PS
Quantity 2.5 g
Synonym Sodium Diethyl Thionophosphate, Ethyl Sodium Phosphorothioate ((EtO)2(NaO)PS) (7CI), Ethyl Sodium Phosphorothioate (6CI), Phosphorothioic Acid, O,O-Diethyl Ester, Sodium Salt (8CI,9CI), O,O-Diethyl S-Sodium Phosphorothioate, O,O-Diethyl Phosphorothioate S-Sodium Salt, Sodium O,O-Diethyl Phosphorothioate
InChI Formula InChI=1S/C4H11O3PS.Na/c1-3-6-8(5,9)7-4-2;/h3-4H2,1-2H3,(H,5,9);/q;+1/p-1
SMILES CCOP(=S)(O[Na])OCC
IUPAC Name diethoxyphosphinothioyloxysodium
Molecular Weight (g/mol) 192.15
Formula Weight 191.999
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.