missing translation for 'onlineSavingsMsg'
Learn More

O,O-Diethyl Dithiophosphate-13C4 Ammonium Salt, TRC

Product Code. 30379905
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30379905 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30379905 Supplier Toronto Research Chemicals Supplier No. TRCD44426710MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O,O-Diethyl Dithiophosphate-13C4 Ammonium Salt
CAS 1329641-22-6
Molecular Formula 13C4H14NO2PS2
Quantity 10 mg
Synonym Phosphorodithioic Acid O,O-Diethyl Ester-13C4 Ammonium Salt, Ammonium Ethyl Phosphorodithioate-13C4, Phosphorodithioic Acid O,O-Diethyl Ester-13C4 Ammonium Salt, Ammonium O,O-Diethyl Dithiophosphate-13C4, Ammonium O,O-Diethyl Phosphorodithioate-13C4, Ammonium Di-O-ethyl Dithiophosphate-13C4, Ammonium Diethyldithiophosphate-13C4, NF 133-13C4, O,O-Diethyl Ammonium Phosphorodithioate-13C4, O,O-Diethylphosphorodithioic Acid-13C4 Ammonium Salt
InChI Formula InChI=1S/C4H11O2PS2.H3N/c1-3-5-7(8,9)6-4-2;/h3-4H2,1-2H3,(H,8,9);1H3
SMILES SP(O[13CH2][13CH3])(O[13CH2][13CH3])=S.N
Molecular Weight (g/mol) 207.23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.