Learn More
O-Nitrophenyl 2,3,4,6-Tetra-O-acetyl-Beta-D-galactopyranoside, TRC
Specifications
Specifications
| Chemical Name or Material | O-Nitrophenyl 2,3,4,6-Tetra-O-acetyl-beta-D-galactopyranoside |
| CAS | 3053-17-6 |
| Molecular Formula | C20 H23 N O12 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C20H23NO12/c1-10(22)28-9-16-17(29-11(2)23)18(30-12(3)24)19(31-13(4)25)20(33-16)32-15-8-6-5-7-14(15)21(26)27/h5-8,16-20H,9H2,1-4H3/t16-,17+,18+,19-,20-/m1/s1 |
| SMILES | CC(=O)OC[C@H]1O[C@@H](Oc2ccccc2[N+](=O)[O-])[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@H]1OC(=O)C |
| IUPAC Name | [(2R,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-(2-nitrophenoxy)oxan-2-yl]methyl acetate |
| Molecular Weight (g/mol) | 469.4 |
| Formula Weight | 469.122 |