Learn More
O-Desmethyl Adapalene, TRC
Specifications
Specifications
| Chemical Name or Material | O-Desmethyl Adapalene |
| CAS | 125316-60-1 |
| Molecular Formula | C27 H26 O3 |
| Quantity | 5 mg |
| Synonym | 6-(4-Hydroxy-3-tricyclo[3.3.1.13,7]dec-1-ylphenyl)-2-naphthalenecarboxylic Acid, 6-[3-(1-Adamantyl)-4-hydroxyphenyl]-2-naphthalenecarboxylic Acid, AHPN, CD 437, CD 437/AHPN |
| InChI Formula | InChI=1S/C27H26O3/c28-25-6-5-22(20-1-2-21-11-23(26(29)30)4-3-19(21)10-20)12-24(25)27-13-16-7-17(14-27)9-18(8-16)15-27/h1-6,10-12,16-18,28H,7-9,13-15H2,(H,29,30)/t16-,17+,18-,27- |
| SMILES | OC(=O)c1ccc2cc(ccc2c1)c3ccc(O)c(c3)[C@@]45C[C@@H]6C[C@@H](C[C@@H](C6)C4)C5 |
| IUPAC Name | 6-[3-(1-adamantyl)-4-hydroxyphenyl]naphthalene-2-carboxylic acid |
| Molecular Weight (g/mol) | 398.49 |
| Formula Weight | 398.1882 |