missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Cresol-d7, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.50g
1g
Specifications
Specifications
| Chemical Name or Material | o-Cresol-d7 |
| CAS | 202325-50-6 |
| Molecular Formula | C7 2H7 H O |
| Quantity | 0.5 g |
| InChI Formula | InChI=1 S/C7H8O/c1-6-4-2-3-5-7(6)8/h2-5,8 H,1H3/i1D3,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c(O)c1[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2,3,4,5-tetradeuterio-6-(trideuteriomethyl)phenol |
| Molecular Weight (g/mol) | 115.18 |
| Formula Weight | 115.1015 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction