Learn More
o-Anisidine-d7, TRC
Product Code. 30339391
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30339391 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30339391 Supplier Toronto Research Chemicals Supplier No. TRCA67352250MG
 Restricted Item
Estimated Shipment: 09-10-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material o-Anisidine-d7
CAS 1219803-70-9
Molecular Formula C7 2H7 H2 N O
Quantity 50 mg
Synonym 2-Methoxy-d3-aniline-d4, 2-Methoxy-d3-aniline-3,4,5,6-d4, 1-Amino-2-(methoxybenzene-d7), o-Anisidine-d7, 2-Methoxyaniline-D7 (methoxy-D3-3,4,5,6-D4), 2-Methoxyaniline-D7
InChI Formula InChI=1S/C7H9NO/c1-9-7-5-3-2-4-6(7)8/h2-5H,8H2,1H3/i1D3,2D,3D,4D,5D
SMILES [2H]c1c([2H])c([2H])c(OC([2H])([2H])[2H])c(N)c1[2H]
IUPAC Name 2,3,4,5-tetradeuterio-6-(trideuteriomethoxy)aniline
Molecular Weight (g/mol) 130.2
Formula Weight 130.1124