missing translation for 'onlineSavingsMsg'
Learn More

o-Anisic acid, 99%

Product Code. p-60944 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10307310

Brand: Thermo Scientific Acros 173751000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 579-75-9 | C8H8O3 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 579-75-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002431
InChI Key ILUJQPXNXACGAN-UHFFFAOYSA-N
Synonym o-anisic acid, o-methoxybenzoic acid, 2-anisic acid, o-methylsalicylic acid, benzoic acid, 2-methoxy, salicylic acid methyl ether, 2-methoxy-benzoic acid, 2-methoxybenzoicacid, o-methoxy benzoic acid, ortho-methoxybenzoic acid
PubChem CID 11370
ChEBI CHEBI:421840
IUPAC Name 2-methoxybenzoic acid
SMILES COC1=CC=CC=C1C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 98°C to 102°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula CH3OC6H4CO2H
Quantity 100 g
Beilstein 10, 64
Solubility Information Solubility in water: 15g/L (20°C)
Formula Weight 152.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material o-Anisic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • o-Anisic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.