Learn More
O-(2-Ethylbutyl) Dabigatran Ethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl 3-((2-((4-(N-(2-Ethylbutoxycarbonyl)carbamimidoyl)anilino)methyl)-1-methylbenzimidazole-5-carbonyl)(pyridin-2-yl)amino)propanoate |
| CAS | 1610758-20-7 |
| Molecular Formula | C34 H41 N7 O5 |
| Quantity | 100 mg |
| Synonym | O-(2-Ethylbutyl) Dabigatran Ethyl Ester, β-Alanine, N-[[2-[[[4-[[[(2-ethylbutoxy)carbonyl]amino]iminomethyl]phenyl]amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-, ethyl ester (ACI), N-[[2-[[[4-[[[(2-Ethylbutoxy)carbonyl]amino]iminomethyl]phenyl]amino]methyl]-1-methyl-1H-ben |
| InChI Formula | InChI=1S/C34H41N7O5/c1-5-23(6-2)22-46-34(44)39-32(35)24-11-14-26(15-12-24)37-21-30-38-27-20-25(13-16-28(27)40(30)4)33(43)41(19-17-31(42)45-7-3)29-10-8-9-18-36-29/h8-16,18,20,23,37H,5-7,17,19,21-22H2,1-4H3,(H2,35,39,44) |
| SMILES | CCOC(=O)CCN(C(=O)c1ccc2c(c1)nc(CNc3ccc(cc3)C(=N)NC(=O)OCC(CC)CC)n2C)c4ccccn4 |
| IUPAC Name | ethyl 3-[[2-[[4-[N-(2-ethylbutoxycarbonyl)carbamimidoyl]anilino]methyl]-1-methylbenzimidazole-5-carbonyl]-pyridin-2-ylamino]propanoate |
| Molecular Weight (g/mol) | 627.733 |
| Formula Weight | 627.317 |