Learn More
Brady™ Nylon Ties
Product Code. 11776885
Change view
Click to view available options
Color:
Blue
Orange
Purple
Red
White
Yellow
Unit Size:
1 pack
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11776885 White 1 pack
11796885 Red 1 pack
11706895 Orange 1 pack
11786885 Blue 1 pack
11716895 Yellow 1 pack
11736895 Purple 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11776885 Supplier Brady™ Supplier No. 229955
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
  • Cross-laminated polyethylene
  • Matte finish
Specifications
Type Ties
Material Nylon
Color White
Length (Metric) 203.00 mm
Width (Metric) 4 mm
Dimensions (L x W) 203 x 3.60 mm
Quantity 100/Pk.
For Use With (Application) Ideal for color-coding hard to mark items