Learn More
Brady™ Nylon Ties
Product Code. 11746895
Change view
Click to view available options
Color:
Black
Unit Size:
1 pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11746895 Black 1 pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11746895 Supplier Brady™ Supplier No. 229962
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Type Ties
Material Nylon
Color Black
Width (Metric) 4 mm
Quantity 100/Pk.
For Use With (Application) Ideal for color-coding hard to mark items