Learn More
Masterflex™ Nylon Threaded Reducing Nipples
Description
- Nylon Threaded Reducing Nipples
- Threaded fittings and connections offer a more permanent installation
- Available in a variety of matierials and sizes
- Pack of Six
Specifications
Specifications
| Material | Nylon |
| Overall Length (English) | 1.375 in. |
| Overall Length (Metric) | 34.93 mm |
| Packaging Quantity | 6 Fittings |
| Port Size | 3/4 in. NPT(M) x 3/8 in. NPT(M) |
| Sterilization Method | Ethylene Oxide |
| Thread Size | 3/4 in. NPT(M) |
| Width (Metric) | 29 mm |
| Width (English) | 1.14 in. |
| Product Type | Reducing Nipple |