Learn More
Cole-Parmer™ Nylon Manifold, 6 outlets
Product Code. 11755228
Click to view available options
Tubing Size:
1/2 in.
1/4 in.
Unit Size:
Each
Product Code. 11755228
Brand: Cole-Parmer™ 3152243
Only null left
Add to basket
Add to basket
Expand your flow and line capacity
  • Constructed of nylon for economic general applications.
  • Offers good chemical compatibility.
  • Available in a variety of configurations to suit a wide range of applications.
Specifications
Material Nylon
Overall Length (English) 4.75 in.
Overall Length (Metric) 12.07 cm
Min. Operating Temperature (English) -60°F
Min. Operating Temperature (Metric) -54°C
Tubing Size 1/4 in.
Inlet 1/4 in. NPT(F)
Outlet Port 1/8 in. NPT(F)
Width (Metric) 2.54 cm
Width (English) 1 in.
Height (Metric) 2.54 cm
Height (English) 1 in.
Maximum Pressure 200 psi
Max. Operating Temperature (English) 180°F
Max. Operating Temperature (Metric) 82°C
Product Type Nylon Manifold, 6 Outlets