Learn More
Masterflex™ Nylon Male Threaded Adapter Barbed Fitting
Click to view available options
For Use With:
1 x 3/4 in. tubing I.D.
1/8 x 3/16 in. tubing I.D.
Unit Size:
Pack of 10
Description
- No leaks, flow restrictions or costly downtime
- A crucial component in setting up a fluid handling system
- Typically holds pressures up to 30 psi (2 bar)
- Slip flexible tubing or hose onto the barb to create a secure seal
Specifications
Specifications
| Material | Nylon |
| For Use With | 1 x 3/4 in. tubing I.D. |
| Packaging Quantity | 10 Pack |
| Sterilization Method | Ethylene Oxide |
| Shape | Straight |
| Maximum Pressure | 30 psi (2 bar) |
| Product Type | Barbed Fitting |