Learn More
Brady™ Nylon Cloth Labels for Portable Label Printers
Product Code. 17673205 Shop All Brady Products
Change view
Click to view available options
Height (Metric):
12.7 mm
19 mm
Width (Metric):
19 mm
25.4 mm
38.1 mm
50.8 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Width (Metric) Unit Size
17673205 19 mm 50.8 mm Each
17633205 12.7 mm 19 mm Each
17643205 12.7 mm 25.4 mm Each
17663205 19 mm 38.1 mm Each
17653205 19 mm 25.4 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17673205 Supplier Brady™ Supplier No. 152094
  • ONLINE EXCLUSIVE PRICE
    40.20€
    33.95€ / Pack of 1
    Save 6.25€
    15% Off
View all promotions
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
Autoclavable nylon cloth labels for portable label printers, suitable for low temperatures.
Nylon cloth labels for Brady™ M210 and M211 portable label printers.
  • Aggressive, permanent adhesive
  • Die-cut labels
  • For low temperatures, such as in freezers or liquid nitrogen
  • Autoclavable
Specifications
Product Type Label
Color Black on White
For Use With (Equipment) Brady M210 Label Printer, Brady M210-LAB Label Printer, Brady M211 Label Printer
Material Nylon Cloth
Print Technology Thermal Transfer
Width (English) 2 in.
Width (Metric) 50.8 mm
Height (English) 0.75 in.
Height (Metric) 19 mm
Thickness (English) 0.0065 in
Thickness (Metric) 0.165 mm
Dimensions (L x W) 50.80 mm (W) x 19.05 mm (H)
Adhesive Type Permanent Acrylic
Autoclavable Autoclavable
Sterility Non-sterile
Finish Matte
Format Cartridge, Label
Temperature Range (English) -40°F to +194°F
Temperature Range (Metric) -40°C to +90°C
Certifications/Compliance 2011/65/EU and 2015/863
For Use With (Application) Component and Equipment Labeling, Electrical Labeling, General Identification, Laboratory Labeling
Quantity 73
Alternate Part Number M21-137-499