Learn More
Masterflex™ Nylon Barbed NPT Female Pipe Adapters
Product Code. 11790139
Click to view available options
Material:
Nylon
Unit Size:
Pack of 25
Product Code. 11790139
Brand: Masterflex™ 3150040
Only null left
Add to basket
Add to basket
Designed for quick and easy assembly without tools
  • Combine these simple and economical fittings with flexible tubing.
  • For use in low-pressure applications (generally less than 30psi).
Specifications
Description Barb x NPT(F) Connection
Material Nylon
Sterilization Method Ethylene Oxide
Width (Metric) 16 mm
Product Type Barbed NPT female pipe adapters, Nylon, 1//8 in. NPT x 1/8 in. ID, 1/16 in., 1-1/32 in., 5/8 in.