missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NVS MLLT-1
GREENER_CHOICE

Product Code. p-7250138
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18767256

Brand: Tocris Bioscience™ 7482/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective MLLT1/3 inhibitor

NVS-MLLT-1 is a selective inhibitor of YEATS domain of MLLT1 and MLLT3 (IC50 values are 150 and 254 nM for MLLT1 and MLLT3, respectively). It shows selectivity for YEATS2/4 domains and over all bromodomains (CERC2 IC50 > 40 μM).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[2-[[(2 S)-2-Methyl-1-pyrrolidinyl]methyl]-1 H-benzimidazol-6-yl]-6-isoquinolinecarboxamide
CAS 2561471-13-2
Quantity 10 mg
Target Bromodomain Inhibitors
Molecular Formula C23H23N5O
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.