missing translation for 'onlineSavingsMsg'
Learn More

Nutlin-3, Thermo Scientific™

Product Code. 17595575 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 17595575

Brand: Thermo Scientific Acros 469201000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 548472-68-0
Quantity 100 mg
Molecular Formula C30 H30 Cl2 N4 O4
Formula Weight 581.5
Physical Form Crystalline Powder
Color White to Light-yellow
Purity ≥97.5%
InChI Key BDUHCSBCVGXTJM-UHFFFAOYNA-N
SMILES COC1=CC=C(C2=NC(C(N2C(=O)N2CCNC(=O)C2)C2=CC=C(Cl)C=C2)C2=CC=C(Cl)C=C2)C(OC(C)C)=C1
IUPAC Name 4-[4,5-bis(4-chlorophenyl)-2-[4-methoxy-2-(propan-2-yloxy)phenyl]-4,5-dihydro-1H-imidazole-1-carbonyl]piperazin-2-one
Molecular Weight (g/mol) 581.49
Chemical Name or Material Nutlin-3
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.