missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies (±)-Nutlin-3

Product Code. 17120272
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 17120272

Brand: STEMCELL Technologies 73752

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

p53 pathway activator; Inhibits MDM2

  • (±)-Nutlin-3 is a small-imidazoline-based mouse double minute 2 (MDM2) protein antagonist which disrupts MDM2-p53 interaction (IC50 = 0.09 μM; Vassilev et al.) MDM2 binds the p53 tumor suppressor protein with high affinity and negatively modulates its transcriptional activity and stability
  • By disrupting this interaction, (±)-Nutlin-3 therefore promotes the expression of p53-regulated genes and exhibits potent antiproliferative activity in cells with functional p53, but not in cells with mutated p53 (Vassilev et al.), Gu et al.)

Specifications

CAS 548472-68-0
Quantity 1 mg
Molecular Formula C30H30Cl2N4O4
Purity >98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.