Learn More
Thermo Scientific™ Nunc™ Polycarbonate Cell Culture Inserts in Multi-Well Plates
missing translation for 'orderingAttributeHoverText'
Pore Size:
0.4 μm
3 μm
8 μm
Dish Type:
12-Well Plate
24-Well Plate
6-Well Plate
Culture Area:
0.47 cm2
1.13 cm2
3.14 cm2
4.1 cm2
Förpackningsstorlek
Pack of 24
Pack of 48
Beskrivelse
Cultivate most cell types without matrix coating when using Thermo Scientific™ Nunc™ Polycarbonate Membrane Inserts in Multidishes.
Features:
- Thermo Scientific™ Nunclon™ Delta-treated and tested for cell attachment
- Tabs provide better grip and easy handling
- Nontoxic and nonpyrogenic
Ordering Information:
Packed in resealable bags
Tekniske data
Tekniske data
| Description | Polycarbonate Cell Culture Inserts with 3 micron pore size in 12-well plate |
| Inserts per Plate | 12 |
| Culture Area | 1.13 cm2 |
| Material | Polycarbonate (Membrane) |
| Volume (Metric) Working | 1.1 mL |
| Pore Density | <1.7 x 106 pores/cm2 |
| Pore Size | 3 μm |
| Type | Culture Insert |
| For Use With | Multidish 12 |
| No. per Pack | 12 |
| Vis mere |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion