Learn More
Thermo Scientific™ Nunc™ Low Cell Binding Surface Dishes and Multidishes
Click to view available options
Volume (Metric) Working:
1 mL
2 mL
3 mL
Unit Size:
Pack of 7
Description
Prevent attachment of adherent cells to the surface, allowing formation of anchorage-independent cell colonies with Thermo Scientific™ Nunc™ Low Cell Binding Surface Dishes and Multidishes.
- Prevents attachment of adherent cells to the surface
- Ideal for culturing embryroid bodies, neurospheres or other anchorage-independent cell clusters
- Sterilized by gamma-irradiation and certified to be non-pyrogenic
Includes: Lid
Specifications
Specifications
| Closure Included | Closure Included |
| Closure Material | Polystyrene |
| Description | Multidish |
| Bottom Shape | Flat |
| Capacity (Metric) | 3 mL (Well) |
| Material | Polystyrene |
| Sterility | Sterile |
| Surface Treatment | Nunclon Delta |
| Type | Multi Dish |
| Volume (Metric) Working | 3 mL |
| Show More |