Learn More
Thermo Scientific™ Nunc™ Lab-Tek™ II Chambered Coverglass
Product Code. 13013053
Click to view available options
Wells:
1
2
4
8
Material:
Borosilicate Glass (1.5)
Description:
1-well Chambered Coverglass w/ non-removable well
2-well Chambered Coverglass w/ non-removable wells
4-well Chambered Coverglass w/ non-removable wells
8-well Chambered Coverglass w/ non-removable wells
Unit Size:
Pack of 16
Pack of 96
Product Code. 13013053
Brand: Thermo Scientific™ 155379
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Culture and view living cells on a single device. This coverglass has optimum optical characteristics for high-magnification microscopy and confocal image analysis.
Culture, examine, and analyze living cells on a single device. Thermo Scientific™ Nunc™ Lab-Tek™ II Chambered Coverglass with a No. 1.5 borosilicate glass bottom offers the ideal optical characteristics required for high-magnification microscopy and confocal image analysis. Available in 1-, 2-, 4-, and 8-well formats. The upper chamber structure cannot be removed from the coverglass.
  • Non-removable polystyrene chambered media chamber securely mounted to extra-thin (No. 1.5) borosilicate coverglass for optimum high-power inverted microscopic viewing
  • Gas exchange achieved with minimal evaporation through tabbed polystyrene cover
  • For culturing and viewing living cells
  • Sterile
Ordering Information:
Shipped sterile in convenient stackable incubation trays.
TRUSTED_SUSTAINABILITY
Specifications
Wells 2
Culture Area 4.0 cm2
Lid Material Polystyrene
Description 2-well Chambered Coverglass w/ non-removable wells
Surface Treatment Cell Culture
Material Borosilicate Glass (1.5)
Volume (Metric) Working 1.0 to 2.0 mL
Thickness (Metric) 0.16 to 0.19 mm
Includes Clear polystyrene lid with handles on the top and side for aseptic manipulations
No. per Pack 16
Dimensions (LxW) 55 x 24 mm
Sterility Sterile
Closure Color Clear
Show More Show Less
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.