Learn More
Thermo Scientific™ Nunc™ Lab-Tek™ II Chamber Slide™ System
Product Code. 11387704
Click to view available options
Wells:
1
2
4
8
Unit Size:
Pack of 16
Pack of 96
Product Code. 11387704
Brand: Thermo Scientific™ 154526
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Grow cells directly on slides with the Nunc Lab-Tek II Chamber Slide System.
Grow, stain, and examine cells directly on the Thermo Scientific Nunc Lab-Tek II Chamber Slide, which consists of a removable polystyrene media chamber attached to a standard glass slide. The slide surface is specially treated for adherent cell culture. Available in 1-, 2-, 4-, and 8-well formats.
Features
  • Slide surface that is specially treated with a unique RS wash to ensure consistent cell growth
  • Tabbed polystyrene lid that allows gas exchange with minimal evaporation
  • Removal tool that separates media chamber from glass slide
  • Biocompatible adhesive hydrophobic well border printed on slide
  • White frosted writing area
Includes: chamber removal tool
Additional information
  • Can be purchased in case of 96 or pack of 16
  • Shipped sterile in convenient stackable incubation trays
Warranty: 90 days
TRUSTED_SUSTAINABILITY
Specifications
Wells 4
Culture Area 1.7 cm2
Lid Material Polystyrene
Description 4-well Chamber Slide w/ removable wells
Surface Treatment Cell Culture
Material RS Glass
Volume (Metric) Working 0.5 to 1.0 mL
Includes Clear polystyrene lid with handles on the top and side for aseptic manipulations
No. per Pack 16
Dimensions (LxW) 74 x 25 mm
Sterility Sterile
Closure Color Clear
Quantity 1 Ea.
Show More Show Less
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.