Learn More
Thermo Scientific™ Nunc™ Cell Factory™ System Accessories
Product Code. 10274451
Click to view available options
Product Type:
Adapter Closure
Airvent Filter
Airvent Filter, 0.22 μm
Cover Cap
FEP Connector
Filter Adapter Cap, Sterile
Filter Assembly, 200 cm⊃2
Filter Assembly, 37 mm
HDPE Connector
PC Connector
System Start-Up Kit
Tubing Set, Female MPC
Tubing Set, Male MPC
Vent Cap
Vent/Close Cap
For Use With (Equipment):
Cell Factory Systems (All)
EasyFill Cell Factory Systems
Unit Size:
Each
Pack of 10
Pack of 100
Pack of 12
Pack of 2
Pack of 20
Pack of 40
Product Code. 10274451
Brand: Thermo Scientific™ 170769
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories.
Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories. Reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the Cell Factory system.
Includes:
  • HDPE connector
  • Two cover caps
  • Two blue sealing caps
  • No. 4210 bacterial air vent filter
  • Tubing and clamp
All items are sterile.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifications
Description Cell Factory System Start-up Kit
For Use With (Equipment) Cell Factory Systems (All)
Includes
  • HDPE connector
  • Two cover caps
  • Two blue sealing caps
  • No. 4210 bacterial air vent filter
  • Tubing and clamp
All items are sterile.
No. per Case 1
No. per Pack 1
Product Type System Start-Up Kit
Bottom Shape Flat
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.