Learn More
Thermo Scientific™ Nunc™ 1.8mL Externally-Threaded Universal Tubes with 1D (linear) code on the side and 2D code on tube bottom, not racked
GREENER_CHOICE
Use this versatile tube either inside a large storage system, or in smaller scale when used for traditional cryostorage in dewars.
Brand:  Thermo Scientific™ 374502NOV
Product Code. 15415004
  • POA / Pack of 480
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
Use versatile Thermo Scientific™ Nunc™ 1.8 mL Externally-threaded Universal Tubes either inside a large storage system, or in smaller scale when used for traditional cryostorage in dewars. Available racked or in bulk, with 2D barcodes or non-coded.
Scalable
  • Tubes and rack are ideal for both lower-throughput and high-throughput use
  • Feature graduations and white patch for manual marking
  • Can be used in traditional cyrostorage boxes made for 10x10 arrays
  • External threads limit contamination
  • Automation-pickable in Universal Latch Rack
Innovative Universal Cap Design
  • Single-piece construction with molded-in gasket
  • Cannot be over-tightened manually or with automated capping tools
  • Thermoplastic elastomer gasket material is compatible with storage at room temperature down to vapor phase of liquid nitrogen
Secure Tracking
  • Permanently-bonded,unique 2D barcode laser-etched onto the base of tube to securely identify and track samples
  • 2D barcode readers instantly scans each tube's 2D barcode, registering the tube in any application or database, or integrates via several export options
  • Scan the tube to access sample information and identify the contents
Stringent Quality Control
  • Every 2D barcoded storage tube is scanned to ensure readability
  • Barcodes are checked against database of previously-assigned codes to prevent duplicates
  • Every tube has a Sterility Assurance Level of 10superscript 6 and is made with class VI medical grade resins