Learn More
Macherey-Nagel™ NucleoVac 24 Vacuum Manifold
Product Code. 15543616
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15543616 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15543616 Supplier Macherey-Nagel™ Supplier No. 740299
  • ONLINE EXCLUSIVE PRICE
    295.00€
    262.50€ / Pack of 1
    Save 32.50€
    11% Off
In Stock
Add to basket
Add to basket
Vacuum manifold for processing 1–24 NucleoSpin or NucleoSnap column.
The NucleoVac 24 Vacuum Manifold allows the processing of silica columns for nucleic acid purification by vacuum.
  • Silica columns (NucleoSpin™ and NucleoSnap) can be attached to the upper luer slots of the manifold
  • Binding and washing steps are carried out by vacuum instead of centrifugation
  • Parallel processing of silica columns saves signficant time
  • Enables fast processing of large sample volumes
  • Polypropylene
Specifications
Product Type Vacuum Manifold
Format 24 Samples
Capacity ≤900 mL (Waste)
Description Polypropylene
Content And Storage 1 Ea., 15°C to 25°C
For Use With (Equipment) NucleoSpin or NucleoSnap columns
Includes Vacuum Manifold, 24 Mini Adapters, 24 Luer plugs, 2 tubing connections, 1 closing plug
Ports Luer openings
Quantity 1 Ea.