missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoTrap suspension for gel extraction

Product Code. 10796712
This item is not returnable. View return policy

Product Code. 10796712

Brand: Macherey-Nagel™ 740589

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The silica matrix based suspension for gel extraction without columns.

NucleoTrap™ is a silica matrix based kit for isolation of highly pure DNA fragments from agarose gels and aqueous solutions (desalination). The kit can also be used for the removal of nucleotides and/or biotin after labelling reactions as well as for the removal of enzymes.

By means of a specially activated spherical silica matrix it is possible to extract DNA fragments from agarose gels or solvents in a very mild manner, without the use of organic solvents. Nucleic acids are bound to the NucleoTrap™ matrix in the presence of a chaotropic salt. The hydrate shell of the nucleic acids is destroyed by this binding buffer, thus facilitating adsorption to the specially treated NucleoTrap™ matrix. A washing step is followed by the elution of DNA with low-salt elution buffer or distilled water.Kit components: NucleoTrap™ suspension, buffers.
  • Silica matrix technology
  • Fragment size: 20bp to 20kb
  • Sample material: 20mg to 600mg agarose gel
  • Elution volume: 20μL to 50μL
  • Binding capacity: 6μg/10μL suspension
  • Typical Recovery: 50% to 90%
  • Preparation time: 60 min./6 preps
  • Format: silica bead suspension
For extraction of DNA from agarose gels, suitable, for example, for subsequent cloning sequencing, PCR, restriction analysis. Support protocol for concentration and desalination of reaction mixtures.

Specifications

Capacity 0.6 μg/10 μL suspension
Isolation Technology Silica Matrix Technology
For Use With (Application) For Extraction of DNA From Agarose Gels
No. of Reactions 100
Product Type NucleoTrap™ Suspension
Sample Volume <200 mg
Test Time 60min.
Form Silica Bead Suspension
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.