missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ 96 RNA Tissue plate, 96‐well kit for RNA purification

Product Code. 16371616
Change view
Click to view available options
Quantity:
4 x 96 Preps
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16371616 4 x 96 Preps Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16371616 Supplier Macherey-Nagel™ Supplier No. 740733.4S
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

RNA isolation in flexible 96‐well plate format

  • Proven 96-well format for high throughput isolation of RNA
  • Time-saving isolation of RNA
  • Processing under vacuum or by centrifugation
  • Suitable for manual and automated processing

Specifications

Concentration 50 to 200 ng/μL
Format 96-well Plate
Isolation Technology Silica Membrane
Sample Type Cells, Tissue
For Use With (Equipment) Vacuum or Centrifuge
For Use With (Application) RNA Purification
No. of Reactions 4 x 96
Purity A260/A280: 1.90–2.10
Quantity 4 x 96 Preps
Sample Size < 107 cultured cells (centrifugation), < 30 mg tissue (centrifugation), saliva (collected with Oragene™)
Size >200 nt
Test Time 45 min/ 6 strips
Yield 20 μg (2 x 106 HeLa cells), 20 μg (20 mg mouse liver)
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.