Learn More
Macherey-Nagel™ NucleoSpin™ RNA Clean-up XS, Micro kit for RNA clean up and concentration
Product Code. 11912482
Click to view available options
No. of Reactions:
10
50
250
Unit Size:
10 reactions
250 reactions
50 reactions
Product Code. 11912482
Brand: Macherey-Nagel™ 740903.10
Highly efficient clean up and concentration of RNA samples.
Specifications
Detection Method One-phase extraction
Isolation Technology One-phase Extraction
Final Product Type NucleoSpin™ RNA Clean-up XS Column
For Use With (Application) For RNA isolation from cultured cells, animal tissue, plant tissue, microorganisms
Includes Collection Tubes, Collection Tubes (2 mL), Collection Tubes (1.5 mL), buffers
No. of Reactions 10
Product Type NucleoSpin™ RNA Clean-up XS Column
Quantity 1 Each
Sample Size <300 μL solution containing <90 μg RNA
Form Reagent
compliance-icons
Product Identifier
  • NucleoSpin RNA Clean-up XS
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
Supplemental information
  • MIXTURE LIST-Contains : guanidinium thiocyanate