Learn More
Macherey-Nagel™ NucleoSpin™ miRNA Plasma Kit
Specifications
Specifications
| Format | Mini Spin Column |
| For Use With (Application) | Isolation of small RNA from plasma or serum for cancer research |
| Includes | NucleoSpin™ miRNA Columns, Collection Tubes (2 mL), buffers, RNase-free rDNase |
| No. of Reactions | 10 Reactions |
| Product Type | NucleoSpin™ miRNA Plasma Kit |
| RNase | Yes |
| Size | <1000 nt / bp |
| Test Time | 40 min./10 Reaction and 70 min./10 Reaction |
Safety and Handling
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1